missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-amino-5-bromobenzoate, 98%, Thermo Scientific Chemicals Product Code.: 11442648

Methyl 2-amino-5-bromobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11442648
25 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11442648

Brand: Thermo Scientific Alfa Aesar A16666.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

Solubility
Soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents, strong reducing agents, acid halides, acid anhydrides, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52727-57-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00144761
InChI Key QVNYNHCNNGKULA-UHFFFAOYSA-N
Synonym methyl 5-bromoanthranilate, 2-amino-5-bromobenzoic acid methyl ester, 5-bromoanthranilic acid methyl ester, 2-amino-5-bromo-benzoic acid methyl ester, methyl 5-bromo-2-aminobenzoate, benzoic acid, 2-amino-5-bromo-, methyl ester, benzoic acid, 2-amino-5-bromo-, methyl ester, hydrochloride, pubchem3344, methyl-5-bromoanthranilate, acmc-1ats7Show MoreShow Less
PubChem CID 736224
IUPAC Name methyl 2-amino-5-bromobenzoate
SMILES COC(=O)C1=CC(Br)=CC=C1N

Specifications

Melting Point 71°C to 73°C
Quantity 25 g
Beilstein 972720
Solubility Information Soluble in water.
Formula Weight 230.06
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-bromobenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10212566 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog NumberA16666.14,A16666.06,A16666.22
Lot Number10237657 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16666.14,A16666.06,A16666.22
Lot NumberH5517C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16666.14,A16666.06,A16666.22
Lot Number10134837 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16666.14,A16666.06,A16666.22
Lot Number10185254 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16666.14,A16666.06,A16666.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-amino-5-bromobenzoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.