missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorocarbonylbenzoate, 90%, tech., Thermo Scientific Chemicals

44.00 € - 145.00 €

Chemical Identifiers

CAS 7377-26-6
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.61
MDL Number MFCD00059486
InChI Key CVXXHXPNTZBZEL-UHFFFAOYSA-N
Synonym methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
PubChem CID 81850
IUPAC Name methyl 4-carbonochloridoylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10792602
View Documents Promotion Details
Thermo Scientific Acros
424710010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10472985
View Documents Promotion Details
Thermo Scientific Acros
424710050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7377-26-6
198.61
CVXXHXPNTZBZEL-UHFFFAOYSA-N
81850
COC(=O)C1=CC=C(C=C1)C(=O)Cl
C9H7ClO3
MFCD00059486
methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
methyl 4-carbonochloridoylbenzoate

Specifications

7377-26-6
White to Yellow
90%
H3COOCC6H4COCl
methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
CVXXHXPNTZBZEL-UHFFFAOYSA-N
methyl 4-carbonochloridoylbenzoate
81850
90%
Crystalline Powder
50°C to 59°C
Authentic
C9H7ClO3
MFCD00059486
Solubility in water: hydrolysis
COC(=O)C1=CC=C(C=C1)C(=O)Cl
198.61
198.61
Technical
Methyl 4-chlorocarbonylbenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 230-937-2

Product Identifier
  • Methyl 4-chlorocarbonylbenzoate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only