missing translation for 'onlineSavingsMsg'
Learn More
Methyl anthranilate, 99%, Thermo Scientific Chemicals Product Code.: 11467627

Methyl anthranilate, 99%, Thermo Scientific Chemicals

Product Code. 11467627
250 g
Click to view available options
Quantity:
250 g
500 g
1000 g
This item is not returnable. View return policy

Product Code. 11467627

Brand: Thermo Scientific Alfa Aesar A15681.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-20-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
InChI Key VAMXMNNIEUEQDV-UHFFFAOYSA-N
PubChem CID 8635
IUPAC Name methyl 2-aminobenzoate
SMILES COC(=O)C1=CC=CC=C1N

Specifications

Melting Point 24°C
Boiling Point 260°C
Quantity 250 g
Merck Index 14,6020
InChI Formula 1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl anthranilate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250833 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15681.36,A15681.0B,A15681.30
Lot Number10215330 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog NumberA15681.36,A15681.0B,A15681.30
Lot Number10244438 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog NumberA15681.36,A15681.0B,A15681.30
Lot Number10141291 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA15681.36,A15681.0B,A15681.30
Lot NumberW02B042 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15681.36,A15681.0B,A15681.30
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl anthranilate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.