missing translation for 'onlineSavingsMsg'
Learn More
Methyl carbamate, 99%, Thermo Scientific Chemicals Product Code.: 10451481

Methyl carbamate, 99%, Thermo Scientific Chemicals

Product Code. 10451481
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10451481

Brand: Thermo Scientific Acros 269310050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-55-0
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00007964
InChI Key GTCAXTIRRLKXRU-UHFFFAOYSA-N
Synonym urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czechShow MoreShow Less
PubChem CID 11722
ChEBI CHEBI:76606
IUPAC Name methyl carbamate
SMILES COC(N)=O

Specifications

Melting Point 54.0°C to 56.0°C
Boiling Point 176.0°C to 177.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCO2CH3
Beilstein 03, 21
Merck Index 15, 6110
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: very soluble in ethanol,soluble in acetone and ether
Quantity 5 g
Flash Point 93°C
Formula Weight 75.06
Percent Purity 99%
Physical Form Adhering Crystals
Chemical Name or Material Methyl carbamate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0315852 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number269310010,269312500,269310050
Lot NumberA0424961 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number269310010,269312500,269310050
Lot NumberA0394674 Certificate TypeCertificate of Analysis Date12/03/2021 Catalog Number269310010,269312500,269310050
Lot NumberA0390620 Certificate TypeCertificate of Analysis Date09/11/2020 Catalog Number269310010,269312500,269310050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl carbamate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.