missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl linolenate, 99%, Thermo Scientific Chemicals
159.00 € - 497.00 €
Chemical Identifiers
CAS | 301-00-8 |
---|---|
Molecular Formula | C19H32O2 |
Molecular Weight (g/mol) | 292.46 |
MDL Number | MFCD00135851 |
InChI Key | DVWSXZIHSUZZKJ-YSTUJMKBSA-N |
Synonym | methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate |
PubChem CID | 5319706 |
IUPAC Name | methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate |
SMILES | CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10222532
|
Thermo Scientific Acros
368100010 |
1 g | Glass bottle |
159.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10079001
|
Thermo Scientific Acros
368100050 |
5 g | Glass bottle |
497.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
301-00-8 | |
292.46 | |
DVWSXZIHSUZZKJ-YSTUJMKBSA-N | |
5319706 | |
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC |
C19H32O2 | |
MFCD00135851 | |
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate | |
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate |
Specifications
301-00-8 | |
0.8950g/mL | |
>110°C | |
98.5% min. (GC) | |
CH3(CH2CH=CH)3(CH2)7CO2CH3 | |
MFCD00135851 | |
0.895 | |
Solubility in water: insoluble | |
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC | |
292.46 | |
292.46 | |
Liquid |
Colorless | |
182.0°C (3.0 mmHg) | |
Authentic | |
C19H32O2 | |
1.4700 to 1.4720 | |
02, IV, 1782 | |
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate | |
DVWSXZIHSUZZKJ-YSTUJMKBSA-N | |
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate | |
5319706 | |
99% | |
Methyl linolenate, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 206-102-3
RUO – Research Use Only