missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, Reag. Ph. Eur., Honeywell Fluka™
Indicator, Reag. Ph. Eur.
89.73 € - 875.18 €
Chemical Identifiers
CAS | 547-58-0 |
---|---|
Molecular Formula | C14H14N3NaO3S |
Molecular Weight (g/mol) | 327.334 |
MDL Number | MFCD00007502 |
InChI Key | STZCRXQWRGQSJD-UHFFFAOYSA-M |
Synonym | Helianthin |
PubChem CID | 23673835 |
IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15653820
|
Honeywell-Fluka
32624-100G |
100 g | Glass Bottle | N/A | |||||
15663820
|
Honeywell-Fluka
32624-250G |
250 g | Glass Bottle | N/A | |||||
15643820
|
Honeywell-Fluka
32624-1.5KG |
1.5 kg | Glass Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
547-58-0 | |
327.334 | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
23673835 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
C14H14N3NaO3S | |
MFCD00007502 | |
Helianthin | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
Specifications
547-58-0 | |
MFCD00007502 | |
4732884 | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
23673835 | |
Complies for sensitivity | |
Orange | |
Crystalline Powder |
C14H14N3NaO3S | |
UN3143 | |
Helianthin | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
327.334 | |
327.33g/mol | |
3.0 to 4.4 | |
3.0 to 4.4 (pink to yellow) | |
Methyl Orange, Reag. Ph. Eur. |
Safety and Handling
P301 + P310
H301
Danger
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'einecsNumber' : 208-925-3
missing translation for 'rtecsNumber' : DB6327000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methyl Orange
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium 4-(4- dimethylaminophenylazo) benzenesulphonate