missing translation for 'onlineSavingsMsg'
Learn More

Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™

C5H12O, CAS Number-1634-04-4, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl tert-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, mtbe, methyl tertiary-butyl ether, methyl-tert-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 5L, 54 deg.C, CHEBI:27642, Colorless

16.80 € - 387.00 €

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number 8812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10263960
View Documents
Fisher Chemical
M/4494/08
500 mL Amber glass bottle
16.80 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10376330
View Documents
Fisher Chemical
M/4494/17
2.5 L Amber glass bottle
68.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10154953
Afficher les documents
Fisher Chemical
M/4494/PB17
2.5 L Plastic Coated Glass bottle
70.30 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10131960
Afficher les documents
Fisher Chemical
M/4494/21
5 L HDPE Plastic Bottle
114.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10428990
Afficher les documents
Fisher Chemical
M/4494/25
25 L Metal can
387.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C5H12O, CAS Number-1634-04-4, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl tert-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, mtbe, methyl tertiary-butyl ether, methyl-tert-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 5L, 54 deg.C, CHEBI:27642, Colorless

Spécification

Identifiants chimiques

1634-04-4
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
2-methoxy-2-methylpropane
C5H12O
8812
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CHEBI:27642
CC(C)(C)OC

Spécification

1634-04-4
Colorless
C5H12O
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CC(C)(C)OC
88.15
CHEBI:27642
268mbar at 20°C
-110°C
54°C
8812
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
15413
88.15g/mol
Liquid
Vidéos
Sécurité et traitement

Sécurité et traitement

Identifiant produit
  • tert-Butyl methyl ether

Mention d'avertissement
  • Danger

Catégorie de danger
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 2

Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.

Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

FDS
Documents

Documents

Product Certifications
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis