Learn More
Metro™ Super Erecta™ Heavy Duty Stainless Steel Cantilever Shelf
Heavy duty stainless steel cantilever shelf is a handy and easy way to expand shelving space in Metro™ Super Erecta™ systems.
Brand:  Metro™ 1260CHC
 View more versions of this product
Product Code. 15996551
  • 175.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Drop mat design creates a retaining ledge around the entire deep shelf
  • For use with Super Erecta™ or HD Super™ shelves
  • Width: 12 in.
Order Info
Order rear posts that are taller than front posts to accommodate this type of shelf.
Specifications
Specifications
Wire Shelf
HD Super™
152.4 cm
30.48 cm
60 x 12 in.
60 in.
12 in.
Chrome