missing translation for 'onlineSavingsMsg'
Learn More

Metro™ High-Density qwikTRAK™ Mobile Unit Kits

Kit provides components for shelving units that fit on tracks for added mobility

528.00 € - 1130.00 €

Specifications

Includes Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Product Type Mobile Unit Kit
View More Specs

Products 4
Product Code Brand For Use With (Equipment) Material Weight (English) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Material Weight (English) Price Quantity & Availability  
15986871
View Documents
Metro™
BTMC
Metro Super Erecta, Super Adjustable Shelving System Steel 25 lb.
528.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
15996871
View Documents
Metro™
BTMK3
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System Steel 25 lb.
892.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
15906881
View Documents
Metro™
BTMS
Metro Super Erecta, Super Adjustable Shelving System Stainless Steel 25 lb.
1130.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
15916881
View Documents
Metro™
BTMX3
Metro MetroMax i and Q Shelving System Polymer 18.5 lb.
964.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
Description

Description

  • For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax iQ™ shelving systems
  • Kit includes four 74 in. (188cm) high mobile posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware

Order Info

Order one kit per mobile system. Shelves are sold separately.

Specifications

Specifications

Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Mobile Unit Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ High-Density qwikTRAK™ Mobile Unit Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.