Learn More
Metro™ High-Density qwikTRAK™ Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
Brand: Metro™ BTMK3
This item is not returnable.
View return policy
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax iQ™ shelving systems
- Kit includes four 74 in. (188cm) high mobile posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per mobile system. Shelves are sold separately.
Specifications
Mobile Unit Kit | |
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System | |
25 lb. | |
Steel |
Metroseal Green Epoxy | |
Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware | |
11.4 kg |
Product Content Correction
Product Title
Metro™ High-Density qwikTRAK™ Mobile Unit Kits > Metroseal 3 with Microban
Spot an opportunity for improvement?Share a Content Correction