missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i Basket Glassware Cart
Level 2 cart with built-in Microban antimicrobial product protection allows arrangement configuration, ensuring storage and movement won't disrupt your workday.
Brand: Metro™ GWBX3-1
This item is not returnable.
View return policy
Description
- Modular design offers ultimate flexibility.
- Reinforced polymers provide superior strength (up to 800 lb. per shelf).
- Corrosion-proof, suitable for damp and humid conditions.
- Lifetime warranty against rust and corrosion.
- Shelves wipe down easily and have a retaining edge that contains drips and spills.
- Two-basket and four-basket capacity glassware solutions are offered in a spacesaving 18 x 36 in. (457 x 914mm) footprint.
- Solid MetroMax bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning.
- Wide range of accessories available separately, including all-polymer casters for maximum corrosion resistance and premium performance.
Order Info
Casters not included (5PC Series Metro casters are recommended for superior lifetime performance).
Specifications
Glassware Cart | |
Taupe, Blue | |
36 x 18 x 4 in. (91.4 x 45.7 x 10.2 cm) | |
40 in. | |
Solid MetroMax i bottom shelf | |
91.4 cm | |
362.8 kg | |
1 | |
45.7 cm |
362.8 kg | |
2-Basket Unit | |
Glassware Transportation | |
101.6 cm | |
36 in. | |
800 lb. | |
Polymer | |
18 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction