Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
Brand:  Metro™ 1860NFS
 View more versions of this product
Product Code. 15916611
  • 479.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Specifications
Solid Shelf
Autoclavable
Metro™ Super Erecta™ Shelving System
152.4 cm
45.7 cm
Silver
60 x 18 in. (152.4 x 45.7 cm)
60 in.
18 in.
Stainless Steel