missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable

Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.

280.00 € - 574.00 €

Specifications

Autoclavable Autoclavable
For Use With (Equipment) Metro™ Super Erecta™ Shelving System
Product Type Solid Shelf
Color Silver
Material Stainless Steel
View More Specs

Products 23
Product Code Brand Width (English) Length (English) Price Quantity & Availability  
Product Code Brand Width (English) Length (English) Price Quantity & Availability  
15966651
View Documents
Metro™
2430NFS
24 in. 30 in.
394.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15976571
View Documents
Metro™
1442NFS
14 in. 42 in.
347.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15946591
View Documents
Metro™
1830NFS
18 in. 30 in.
325.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15936581
View Documents
Metro™
1460NFS
14 in. 60 in.
412.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15926601
View Documents
Metro™
1842NFS
18 in. 42 in.
394.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15916611
View Documents
Metro™
1860NFS
18 in. 60 in.
479.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15976611
View Documents
Metro™
2124NFS
21 in. 24 in.
364.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15916631
View Documents
Metro™
2142NFS
21 in. 42 in.
451.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15966631
View Documents
Metro™
2148NFS
21 in. 48 in.
451.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15966661
View Documents
Metro™
2442NFS
24 in. 42 in.
475.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15986571
View Documents
Metro™
1448NFS
14 in. 48 in.
347.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15996561
View Documents
Metro™
1430NFS
14 in. 30 in.
286.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15966671
View Documents
Metro™
2460NFS
24 in. 60 in.
574.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15916661
View Documents
Metro™
2436NFS
24 in. 36 in.
394.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15926621
View Documents
Metro™
2130NFS
21 in. 30 in.
371.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15986561
View Documents
Metro™
1424NFS
14 in. 24 in.
280.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15936571
View Documents
Metro™
1436NFS
14 in. 36 in.
286.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15986591
View Documents
Metro™
1836NFS
18 in. 36 in.
325.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15956601
View Documents
Metro™
1848NFS
18 in. 48 in.
394.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15996661
View Documents
Metro™
2448NFS
24 in. 48 in.
475.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15906591
View Documents
Metro™
1824NFS
18 in. 24 in.
315.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15976621
View Documents
Metro™
2136NFS
21 in. 36 in.
371.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
15946641
View Documents
Metro™
2160NFS
21 in. 60 in.
564.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
Description

Description

  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications

Specifications

Autoclavable
Solid Shelf
Stainless Steel
Metro™ Super Erecta™ Shelving System
Silver
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.