Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
Brand:  Metro™ 2124NFS
 View more versions of this product
Product Code. 15976611
  • 364.00 € / Each
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Specifications
Solid Shelf
Autoclavable
Metro™ Super Erecta™ Shelving System
61 cm
53 cm
Silver
24 x 21 in. (61 x 53 cm)
24 in.
21 in.
Stainless Steel