Learn More
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits
Connects stationary end units to overhead track sets.
Brand:  Metro™ TTE24K3
 View more versions of this product
Product Code. 15977291
  • 728.00 € / Each
Estimated Shipment: 01-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Kit includes 86 in. (218.5cm) posts and hardware necessary for connecting stationary end units to track
  • Super Erecta Shelves open-wire design minimizes dust accumulation, allows a free circulation of air and greater visibility of stored items.
MetroSeal 3 Finish is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
One kit is requird per single system. Shelves sold separately.
Specifications
Specifications
Stationary End Unit Kit
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
12 lb.
24 in.
61 cm
Steel
Metroseal Green Epoxy
86 in. Posts and connecting hardware
5.4 kg
24 in.
61 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits > Metroseal 3 with Microban; Shelf width: 24 in.