Learn More
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits
Connects stationary end units to overhead track sets.
677.00 € - 1165.00 €
Specifications
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes 86 in. Posts and connecting hardware
Product Type Stationary End Unit Kit
View More Specs
Products 9
Product Code Brand Material Width (English) Shelf Finish Price Quantity & Availability  
Product Code Brand Material Width (English) Shelf Finish Price Quantity & Availability  
15927291
View Documents
Metro™
TTE18S
Stainless Steel 18 in. None
1140.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15957291
View Documents
Metro™
TTE21S
Stainless Steel 21 in. None
1155.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15987291
View Documents
Metro™
TTE24S
Stainless Steel 24 in. None
1165.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15907291
View Documents
Metro™
TTE18C
Steel 18 in. Chrome
677.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15937291
View Documents
Metro™
TTE21C
Steel 21 in. Chrome
687.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15967291
View Documents
Metro™
TTE24C
Steel 24 in. Chrome
708.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15917291
View Documents
Metro™
TTE18K3
Steel 18 in. Metroseal 3 with Microban™
703.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15947291
View Documents
Metro™
TTE21K3
Steel 21 in. Metroseal 3 with Microban™
713.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15977291
View Documents
Metro™
TTE24K3
Steel 24 in. Metroseal Green Epoxy
728.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
Description
Description
  • Kit includes 86 in. (218.5cm) posts and hardware necessary for connecting stationary end units to track
  • Super Erecta Shelves open-wire design minimizes dust accumulation, allows a free circulation of air and greater visibility of stored items.
MetroSeal 3 Finish is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
One kit is requird per single system. Shelves sold separately.
Specifications
Specifications
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
86 in. Posts and connecting hardware
Stationary End Unit Kit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits