Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
Brand:  Metro™ TTM18S
 View more versions of this product
Product Code. 15917301
  • 959.00 € / Each
Estimated Shipment: 01-09-2025
to see stock.
Explore more special offers
Add to basket
Includes:
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Specifications
Mobile Unit Kit
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
32 lb.
18 in.
45.7 cm
Stainless Steel
None
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
14.4 kg
20.25 in.
51.4 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Metro™ Super Erecta™ Top Track Mobile Unit Kits > Stainless Steel; Shelf width: 457mm (18 in.)