Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
451.00 € - 984.00 €
Specifications
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Product Type Mobile Unit Kit
View More Specs
Products 8
Product Code Brand Material Width (English) Shelf Finish Price Quantity & Availability  
Product Code Brand Material Width (English) Shelf Finish Price Quantity & Availability  
15917301
View Documents
Metro™
TTM18S
Stainless Steel 18 in. None
959.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15947301
View Documents
Metro™
TTM21S
Stainless Steel 21 in. None
979.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15977301
View Documents
Metro™
TTM24S
Stainless Steel 24 in. None
984.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15927301
View Documents
Metro™
TTM21C
Steel 21 in. Chrome
451.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15957301
View Documents
Metro™
TTM24C
Steel 24 in. Chrome
466.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15907301
View Documents
Metro™
TTM18K3
Steel 18 in. Metroseal 3 with Microban™
754.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15937301
View Documents
Metro™
TTM21K3
Steel 21 in. Metroseal 3 with Microban™
759.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
15967301
View Documents
Metro™
TTM24K3
Steel 24 in. Metroseal Green Epoxy
774.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
Description
Description
  • Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks.
  • Design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
<
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Specifications
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Mobile Unit Kit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Metro™ Super Erecta™ Top Track Mobile Unit Kits