Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Brand:  Metro™ 2124NS
 View more versions of this product
Product Code. 15996611
  • 273.00 € / Each
Explore more special offers
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
Wire Shelf
21 in.
53 x 63 in. (28 x 28 cm)
Plastic sleeves and wedges
Stainless Steel
530 mm
Metro Super Erecta Shelving System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish > W x L: 53 x 63cm (21 x 28 in.)