missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable
Shelf ledges snap onto shelves and posts for a secure fit without tools to create taller, stackable ledges.
Brand: Metro™ L18N-4S
This item is not returnable.
View return policy
Description
- 4 in. high (10 cm)
- Choice of plated or stainless steel finish
Order Info
Posts and shelves sold separately.
Specifications
Stackable Ledge for Wire Shelving | |
Metro Super Erecta Wire Shelving | |
10 cm | |
45.7 cm |
Stainless Steel | |
4 in. | |
18 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable > Finish: Stainless steel; Height: 4 in. (10cm); Length: 18 in. (45.7cm)
Spot an opportunity for improvement?Share a Content Correction