missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable

Shelf ledges snap onto shelves and posts for a secure fit without tools to create taller, stackable ledges.

31.80 € - 106.00 €

Specifications

Product Type Stackable Ledge for Wire Shelving
For Use With (Equipment) Metro Super Erecta Wire Shelving
Height (English) 4 in.
Height (Metric) 10 cm
View More Specs

Products 5
Product Code Brand Width (English) Material Price Quantity & Availability  
Product Code Brand Width (English) Material Price Quantity & Availability  
15936961
View Documents
Metro™
L36N-4S
36 in. Stainless Steel
71.05 €
Each
Please to purchase this item. Need a web account? Register with us today!
15906961
View Documents
Metro™
L30N-4C
30 in. Chrome
31.80 €
Each
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
 
15916971
View Documents
Metro™
L60N-4S
60 in. Stainless Steel
106.00 €
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
 
15976951
View Documents
Metro™
L24N-4S
24 in. Stainless Steel
52.10 €
Each
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
 
15926951
View Documents
Metro™
L18N-4S
18 in. Stainless Steel
41.85 €
Each
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
 
Description

Description

  • 4 in. high (10 cm)
  • Choice of plated or stainless steel finish

Order Info

Posts and shelves sold separately.

Specifications

Specifications

Stackable Ledge for Wire Shelving
Metro Super Erecta Wire Shelving
4 in.
10 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.