Learn More
Mettler Toledo™ Moisture Analyzer Sample Pans
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Brand:  Mettler Toledo™ 13865
 View more versions of this product
Product Code. 15979405
  • 190.76 € / Pack of 80
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
  • 3.75 in. (96mm) in diameter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
Round
All Mettler Toledo Moisture Analyzers
88.9 mm
Aluminum
Moisture Pan
80 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Moisture Analyzer Sample Pans > For LJ16 Moisture Analyzer; Aluminum