Learn More
BRAND™ Micro-weighing Container Tubes
Includes 10 filters and 3 cover caps
Brand: BRAND™ 708470
38.39 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- One micro-weighing vessel
- For pipettes 0.1-20 μL
- Includes 10 filters and 3 lids
Specifications
Micro-weighing Tube | |
Pipettes 0.1-20 μL | |
One vessel, 10 filters, 3 cover caps |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Micro-weighing Container Tubes > Product Type: Micro-weighing Tube
Spot an opportunity for improvement?Share a Content Correction