missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Sharps Container, Quart
The Sharps Container is easy to use and transport, and provides safe disposal of syringes, needles and other small sharps.
Brand: Heathrow Scientific™ HS120177
This item is not returnable.
View return policy
Description
- Clearly labeled with the Biohazard warning on one side
- Provides safe disposal of syringes, needles and other small sharps
- Stackable to reduce storage space before use
- Clear lid to view fill level
- Slide access lid provides temporary and final closures for safety and security
- Built-in needle removal port, prevents direct contact with sharps
Specifications
1 L | |
Transparent | |
Red | |
19.2 cm | |
HS120177 | |
11.5 cm | |
Container and Lid | |
4.5 in. | |
7.5 in. | |
Sharps Container |
Slide Top | |
1 qt. | |
Reusable | |
11.5 cm | |
Yes | |
Not Autoclavable | |
Sharps | |
4.5 in. | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heathrow Scientific™ Sharps Container, Quart > Sharps Container, Quart; Color: Red; Capacity: 1 L
Spot an opportunity for improvement?Share a Content Correction