missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Santoprene™ Microbore 2-Stop Tube Sets

Choose the size and formulation to suit your flow rate and application needs.

Brand:  Ismatec™ 06431-42

 View more versions of this product

Product Code. 11725159

  • 77.25 € / Pack of 12
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible with Masterflex L/S Multichannel pump head, small cartridges
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Specifications

Specifications

Microbore 2-Stop tube sets, Santoprene; 2.06 mm ID. Pk of 12.
2.06 mm
0.147 in.
1.25 ft.
12 pack
130 mL/min.
0.081 in.
Santoprene
3.76 mm
0.38 m
42
25.3 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers