missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Santoprene™ Microbore 2-Stop Tube Sets
Choose the size and formulation to suit your flow rate and application needs.
Brand: Ismatec™ 06431-12
76.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Compatible with Masterflex L/S Multichannel pump head, small cartridges
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
Specifications
Microbore 2-Stop tube sets, Santoprene; 0.25 mm ID. Pk of 12. | |
0.25 mm | |
1.25 ft. | |
12 pack | |
1.65 mL/min. |
0.0098 in. | |
Santoprene | |
0.38 m | |
12 | |
25.3 psi |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction