missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Santoprene™ Microbore 2-Stop Tube Sets

Choose the size and formulation to suit your flow rate and application needs.

Brand:  Ismatec™ 06431-26

 View more versions of this product

Product Code. 12353760

  • 77.25 € / Pack of 12
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible with Masterflex L/S Multichannel pump head, small cartridges
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Specifications

Specifications

Microbore 2-Stop tube sets, Santoprene; 0.89 mm ID. Pk of 12.
0.89 mm
0.101 in.
1.25 ft.
12 pack
30 mL/min.
0.035 in.
Santoprene
2.59 mm
0.38 m
26
25.3 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.