Learn More
Ismatec™ Platinum-Cured Silicone Microbore 2-Stop Tube Sets
Choose the size and formulation to suit your flow rate and application needs.
Brand:  Ismatec™ 06421-42
121.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11705159
  • 123.00 € / Pack of 6
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compatible with Masterflex L/S Multichannel pump head, small cartridges
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Specifications
Specifications
Microbore 2-Stop tube sets, silicone (platinum-cured); 2.06 mm ID. Pk of 6.
2.06 mm
3.76 mm
0.38 m
130 mL/min.
25.3 psi
0.081 in.
0.147 in.
1.25 ft.
42
3/32 in. Tubing