Learn More
Ansell™ Microchem™ 4000 Apollo Encapsulated Coveralls
Trusted by fire and rescue crews around the world. MICROCHEM™ 4000 APOLLO is a fully encapsulated liquid tight chemical suit designed for use in conjunction with self contained breathing apparatus (SCBA)
1147.95 €
Specifications
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5
Color Green
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate
Ankle Style Elastic, Attached socks, Boot overflaps
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15546454
View Documents
Ansell™
Medium
1147.95 €
Pack of 4
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15556454
View Documents
Ansell™
Large
1147.95 €
Pack of 4
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15566454
View Documents
Ansell™
X-Large
1147.95 €
Pack of 4
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15576454
View Documents
Ansell™
2X-Large
1147.95 €
Pack of 4
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
  • Full Encapsulated coverall
  • Rear double-zip entry
  • Expanded back for internal wearing of self-contained breathing apparatus (SCBA)
  • Rear positioned exhalation valves
  • Internal braces
  • Escape “rip-cord” for rapid doffing in emergency situations
  • Attached socks with boot over flap
  • Attached Ansell Barrier™ gloves with sleeve over flap
Specifications
Specifications
Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5
Fully Encapsulated
Elastic, Attached socks, Boot overflaps
Attached gloves
None
Welded, Taped
Green
Multi-layer Barrier Laminate
Rear-entry double zipper
Hooded
Encapsulated coveralls
Includes Attached Ansell Barrier™ Gloves, integrated socks with boot overflap, fully encapsulated hood