Learn More
Thermo Scientific™ MicroCL 17R Microcentrifuge
GREENER_CHOICE
Quickly perform micro-volume protocols up to 17,000xg, with cooling available.
Brand:  Thermo Scientific™ 75002455
4780.00 EUR valid until 2024-12-31
Use promo code "21629" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 10152694
  • 5975.00 € / Each
Explore more special offers
Description
Description
Ideal for everyday laboratory use, Thermo Scientific MicroCL 17R microcentrifuges provide outstanding processing power, unique safety features and a broad array of microcentrifuge rotors and accessories for microcentrifuge tube processing, with refrigeration available.
Support your micro-volume separation needs with:
  • Performance up to 17,000xg with fast acceleration and deceleration.
  • Refrigerated versions offer cooling from room temperature to set temperature in as little as 9 minutes.
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application:
    • Outstanding corrosion resistance with lightweight engineered polymer rotors.
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids.
    • Eliminate the need for adapters with unique dual row rotor.
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
  • Sleek, lightweight, space-saving design.
  • Durable and easy to clean and maintain.
  • Quiet performance promotes a safe, stress-free environment.
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
17,000 x g
-9°C to +40°C
Benchtop Centrifuge
MicroCL 17R Centrifuge, Refrigerated
2 Years
1 to 99 min. (1 min. increments); HOLD
12.9 x 11.6 x 17.5 in. (33.0 x 29.5 x 44.5 cm)
Thermo Scientific
230 V
13,300 rpm
50 dB(A)
CSA, CE, IVD
24 x 1.5/2.0mL rotor with ClickSeal biocontainment lid
230 V 50/60 Hz
Yes
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
62.0lb. (28.0 kg)
50/60 Hz
24 x 1.5/2 mL Rotor with Clickseal Biocontainment Lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products