Learn More
Ansell™ Microgard™ 1500 Plus FR Coveralls with Hood
MICROGARD™ 1500 Plus FR Coveralls with Hood are made of a highly breathable anti-static SMS fabric which utilises the latest developments in micro fibre technology to ensure good filtration efficiency.
11.23 € - 381.00 €
Specifications
Seam Type Stitched
View More Specs
Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15546574
View Documents Promotion Details
Ansell™
WR17S-00111-03
Medium
381.00 €
Pack of 40
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
15566574
View Documents Promotion Details
Ansell™
WR17S-00111-05
X-Large
381.00 €
Pack of 40
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
15556574
View Documents Promotion Details
Ansell™
WR17S-00111-04
Large
381.00 €
Pack of 40
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
15576574
View Documents Promotion Details
Ansell™
WR17S-00111-06
2X-Large
367.00 €
Pack of 30
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
15586574
View Documents Promotion Details
Ansell™
WR17S-00111-07
3X-Large
367.00 €
Pack of 30
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
17131035
View Documents Promotion Details
Ansell™
WR17S-00111-02
Small
381.00 €
Pack of 40
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
18020130
View Documents Promotion Details
Ansell™
WR17S-00111-08
4X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
18050130
View Documents Promotion Details
Ansell™
WR17S-00111-09
5X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Category III Coverall with 3-piece hood
  • Stitched seams (Red colour)
  • Elasticated wrists, finger loops, waist and ankles.
  • 2-way front zipper with re-sealable storm flap.
  • Flame retardant and anti-static
  • Nonwoven SMMS polypropylene
  • Air and water vapour permeable (“breathable”) to help reduce the risk of heat stress
  • Anti-staticaccording to EN1149-5
  • Optimized body fit ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above).
Specifications
Specifications
Stitched