missing translation for 'onlineSavingsMsg'
Learn More

Contec™ MicroGenesis™ Single Use Microfiber Mop Cover
BIOPHARMA

Mop Cover for use with MicroGenesis 100% synthetic, disposable, microfiber mop.

Brand:  Contec™ MGMP0004

273.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16535314

  • 365.00 € / Pack of 120
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Synthetic microfiber
  • Lightweight and cost effective mop
  • Single use - Av 6 sqm coverage
  • Ideal for the removal of gross soil and fine particulates
TRUSTED_SUSTAINABILITY
Specifications

Specifications

390 x 130 mm
White
Autoclavable
MicroGenesis Mop
Synthetic Microfiber
Mop Head
Non-sterile
Single-use
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Contec™ MicroGenesis™ Single Use Microfiber Mop Cover > Sterile: No

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.