missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Micrograd™ MICROCHEM™ 5000 Coveralls with Hood
Micrograd™ MICROCHEM™ 5000 Coveralls with Hood reaches new levels in chemical protection and has been engineered to protect. The highly visible multi-layer fabric is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams
1225.00 €
Specifications
Product Type | Protective Coveralls |
---|---|
Material | Multi-layer Fabric |
Color | Orange |
Waist Style | Elastic |
Ankle Style | Elastic |
Produktcode | Marke | Size | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Size | Preis | Menge & Verfügbarkeit | |||||
15546474
|
Ansell™
OR50T-00111-03 |
Medium |
1225.00 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15556474
|
Ansell™
OR50T-00111-04 |
Large |
1225.00 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15566474
|
Ansell™
OR50T-00111-05 |
X-Large |
1225.00 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15576474
|
Ansell™
OR50T-00111-06 |
2X-Large |
1225.00 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15586474
|
Ansell™
OR50T-00111-07 |
3X-Large |
1225.00 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
Beschreibung
- Coverall with 2-piece hood
- Elasticated double cuffs (knitted inner cuff), waist and ankles
- Seams are ultrasonically welded and taped
- Highly visible bright orange colour for improved worker safety
- Liquid-tight dual zip designs without the need for additional taping
- Provides a barrier to numerous organic and inorganic chemicals and biological hazards
- Comfortable multi-layer material is lightweight, yet strong and durable
- Protection - >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Anti-static according to EN 1149-5
Spezifikation
Protective Coveralls | |
Orange | |
Elastic | |
Hooded | |
Liquid-tight Double Zipper | |
Cat. III Type 3-B, 4-B, 5-B, EN1073-2, EN14126, EN1149-1 | |
None |
Multi-layer Fabric | |
Elastic | |
Elastic, Knit | |
2-panel, Elastic | |
Taped, Welded | |
Elasticated with double knitted inner cuff | |
Includes Hood |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts