missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Stainless Steel Gel Cleaner
BIOPHARMA

Stainless steel gel cleanser spray, with advanced no-drip technology, for removing rust, oxidation and residue build-up on stainless steel, metal and many other hard surfaces. Available in a 16 oz. spray bottle.

Brand:  Micronova™ SC1-P

109.25 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.



Product Code. 15590713

  • 115.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Clings and glides over most surfaces with minimum effort on contact
  • Apply into seams and crevices, when dealing with rust, oxidation and residue build-up to pre-treat before wiping
  • Nonatomized — will not affect air quality
  • Phosphorus-free
  • Biodegradable surfactants
  • No VOCs
  • NSF A7 rating
Specifications

Specifications

Gel Detergent
Gel
Clear
Store at ambient temperature. Avoid direct sunlight. Protect from freezing.
16 oz.
No
Non-sterile
Clear
Gel Cleaner for Stainless Surface
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : nonylphenol, ethoxylated

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number044911-000 Certificate TypeCertificate of Compliance Date17/09/2024 Catalog NumberSC1-P
Lot Number041898-000 Certificate TypeCertificate of Compliance Date30/05/2024 Catalog NumberSC1-P
Lot Number036003-000 Certificate TypeCertificate of Compliance Date16/01/2023 Catalog NumberSC1-P
Lot Number034641-000 Certificate TypeCertificate of Compliance Date13/12/2022 Catalog NumberSC1-P
Lot Number037550-000 Certificate TypeCertificate of Quality Date18/12/2023 Catalog NumberSC1-P
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ Stainless Steel Gel Cleaner > Stainless Steel Cleaner, gel cleaner, NSF rated, 16 oz. spray bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.