missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ M-Zone™ Bottle Brushes

A range of color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.

Brand:  Micronova™ BRDB-01

47.44 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 12765715

  • 52.10 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

A range of color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.

The Micronova M-Zone™ Bottle Brushes are multipurpose brushes with rounded, stiff polyester bristles and either 12 or 16-inch plastic easy grip handles. Designed to clean bottles and other lab and production equipment.

  • Multipurpose, color-coded plastic bottle brushes
  • Durable, stiff polyester bristles
  • Easy grip plastic handle
  • For versatile and aggressive cleaning of bottles and equipment
  • Made from FDA approved, BPA free base resins
  • Available in Blue, Green, Red, White and Yellow
Specifications

Specifications

Bottle Brush
Plastic
Not Autoclavable
Blue
8.9 cm
Color-Coded Bottle Brushes for Equipment Cleaning
16 in.
40.64 cm
8.9 cm
Polyester
Non-sterile
Keep at ambient temperature
3.5 in.
Reusable
16 in.
40.6 cm
3.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers