missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser
BIOPHARMA

A touch-free, hand care dispensing system that offers hassle-free, wall-mount installation and a gravity-fed pump, for simple bottle replacement and efficient battery use. Bulbs easily attach to soaps and sanitizers for use with the dispenser.

Brand:  Micronova™ ICDISP-3

193.80 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.


Product Code. 11912475

  • 204.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combines touch-free dispensing with no-hassle installation and easy change out of bottles

The M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser is designed for use with a choice of ICCAP Dispensing Capsules that screw onto the corresponding 500 mL replaceable bottles and snap into the well of the dispenser. When the electronic beam is broken by a hand passing under the unit, the bubble on the spout depresses to dispense either 1 or 2 mL of hand care into the palm.

  • Touch-free hand care dispenser
  • Wall mount with drip tray
  • Hassle-free installation and bottle replacement
  • Compact, gravity-fed unit, without the need for tubing
  • Efficient battery use
  • For use with a choice of ICCAP Dispensing Capsules:
    • ICCAP-16 - Green 2 mL dispensing capsule for soaps and sanitizer
    • ICCAP-17IR - Blue 1 mL dispensing capsule for sterile IPA or sanitizer
  • Designed for use with the Micronova hand care 500 mL replacement bottles:
    • Hy-G-Clenz - antibacterial soap, Aquahol - sterile alcohol, NovaClenz - hand sanitizer and LoNa - low sodium hand soap
Specifications

Specifications

Hand Soap Dispenser
Non-sterile
OSHA and CDC
Requires AA Batteries; Requires Handcare 500 mL bottles and ICCAP-16 or ICCAP-17IR
5 in.
Plastic
12 cm
No
Not Autoclavable
White
Pharmaceutical, Biomedical, Healthcare
13 cm
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser > M-Zone MicroDispenser

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.