Learn More
Micronova™ Hand Soaps and Sanitizers

Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
179.00 € - 268.00 €
Specifications
Color-coded | No |
---|---|
Autoclavable | Not Autoclavable |
Container Type | HDPE |
Dimensions (L x W x H) | 7.75 x 1.75 x 3.75 |
Disposable | Single-use |
Product Code | Brand | Product Type | Sterility | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Sterility | Price | Quantity & Availability | ||||||
12822447
![]() |
Micronova™
IC-210 |
Hand Sanitizer | Non-sterile | N/A | N/A | N/A | |||||
13454999
![]() |
Micronova™
IC-210 |
Hand Sanitizer | Non-sterile |
213.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
13127296
![]() |
Micronova™
IC-420 |
Antibacterial Hand Soap | Non-sterile |
190.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12832447
![]() |
Micronova™
IC-220 |
Gloved Hand and Surface Sanitizer | Sterile |
268.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
13107296
![]() |
Micronova™
IC-110 |
Hand and Glove Soap | Non-sterile | N/A | N/A | N/A | |||||
Description
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Hand care products are gentle and effective with built-in skin emollients for frequently washed hands. The 500 mL saddle pump bottles can be used as stand-alone dispensers or with the M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser (ICDISP-3) to ensure uncontaminated results.
- NovaClenz™ Instant Hand Sanitizer, made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including: pseudomonas, salmonella and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
- Hy-G-Clenz™ Antimicrobial Hand Soap, made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
- AquaHol™ Sterile Isopropyl Alcohol Solution, made with 70% Sterile IPA in 30% Wifi Quality Water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
- NovaDyne™ General Purpose Hand Soap, pH balanced gentle lotion hand soap. Made with built-in moisturizers/skin emollients, ideal for frequent hand washing.
- LoNa™ Low Sodium Hand Soap, low ionic, low sudsing hand soap and glove cleanser, specially designed for the high-tech cleanroom; less than 100 ppb sodium. pH balanced and clean enough to be used on the face.
Specifications
No | |
HDPE | |
Single-use | |
19.69 cm | |
9.53 cm | |
500 mL | |
500 mL | |
0.5 kg | |
4.45 cm |
Not Autoclavable | |
7.75 x 1.75 x 3.75 | |
7.75 in. | |
3.75 in. | |
6/Cs. | |
16.9 oz. | |
16.9 oz. | |
1.75 in. |
Safety and Handling
missing translation for 'shelfLife' : 36 Months
- NovaClenz
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.