Learn More
Buerkle™ V4A MicroSampler Head
Buerkle™ V4A MicroSampler Head are interchangeable with different tips and volumes.
Brand:  Buerkle™ 5307-0210
 View more versions of this product
Product Code. 10591731
  • 140.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Interchangeable
  • Allows tips with different volumes to be used
  • Stainless steel AISI 316L
Specifications
Specifications
Heads
Autoclavable
10 mL
25 mm dia. MicroSampler Tube
Stainless Steel (V4A)
Head for MicroSampler
For The Sampling of Small Quantities of Powder