missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Microscope Head, SMZ-171 BH

Stereo microscope head employing the Greenough stereoscopic optical system for large-scale magnification, distortion-free images, and superior depth of field. Motic™ Microscope Head, SMZ-171 BH is a binocular head with a 45° viewing angle.

Brand:  Motic™ 1100200600751

Product Code. 15465502

  • 815.00 € / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Greenough optical system.
  • Binocular head, 45° inclined, 360° rotating.
  • Widefield adjustable eyepieces WF10X/23mm.
  • 6.7:1 Zoom ratio, WD=110mm
Specifications

Specifications

Binocular microscope head
Binocular, 45° inclined, 360° rotating
WF 10X/23 mm, Adjustable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Motic™ Microscope Head, SMZ-171 BH > Binocular microscope head

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.