missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ A-2-MTP Rotor
Compatible with Eppendorf™ 5430/5430 R Centrifuges. Eppendorf™ A-2-MTP Rotor features windshield design for whisper quiet operation.
Brand: Eppendorf™ 5427700005
This item is not returnable.
View return policy
Description
- Max. speed: 2,204 × g (4,680 rpm)
- 2-place swing-bucket rotor for MTP, PCR and Deepwell Plates

Specifications
Microtiter Rotor | |
96 x 0.2 mL | |
2,024 x g | |
4,680 rpm |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ A-2-MTP Rotor > Capacity: 96 x 0.2mL
Spot an opportunity for improvement?Share a Content Correction