Learn More
Fisher BioReagents™ LB Broth, High Salt, Granulated
For propagation and maintenance of E. coli.
Brand:  Fisher BioReagents™ 12801660
 View more versions of this product
Product Code. 12801660
  • POA / 500g
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Material specially granulated to reduce dust and increase solubility rate
  • Sterilize by autoclaving
Specifications
Specifications
Miller Luria-Bertani Broth
Microbiology Media
500 g