Learn More
Milnacipran hydrochloride, MedChemExpress
MedChemExpress Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.
Brand:  Medchem Express HY-B0168A
Encompass_Preferred
 View more versions of this product
Product Code. 16468378
  • 81.00 € / 10mg
Explore more special offers
Description
Description
Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.
Specifications
Specifications
Milnacipran hydrochloride
101152-94-7
10 mg
C15H23ClN2O
H2O : ≥ 100 mg/mL (353.59 mM) ∣DMSO : ≥ 48 mg/mL (169.73 mM)
282.81
99.92%
Solid
White
Research
Neuroscience-Neuromodulation
O=C([C@@]1(C2=CC=CC=C2)[C@@H](CN)C1)N(CC)CC.Cl
282.81
Research