Learn More
Milnacipran hydrochloride, MedChemExpress
MedChemExpress Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.
81.00 € - 336.15 €
Specifications
Chemical Name or Material | Milnacipran hydrochloride |
---|---|
Physical Form | Solid |
CAS | 101152-94-7 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478146
|
Medchem Express
HY-B0168A |
100 mg |
336.15 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16474267
|
Medchem Express
HY-B0168A |
50 mg |
190.35 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468378
|
Medchem Express
HY-B0168A |
10 mg |
81.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.Specifications
Milnacipran hydrochloride | |
101152-94-7 | |
Research | |
Neuroscience-Neuromodulation | |
O=C([C@@]1(C2=CC=CC=C2)[C@@H](CN)C1)N(CC)CC.Cl | |
282.81 | |
Research |
Solid | |
White | |
C15H23ClN2O | |
H2O : ≥ 100 mg/mL (353.59 mM) ∣DMSO : ≥ 48 mg/mL (169.73 mM) | |
282.81 | |
99.92% |
Safety and Handling
H301
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.