Learn More
Fisherbrand™ Mini UV Sterilization Cabinet with Timer
Limits PCR reaction contamination. Fisherbrand™ Mini UV Sterilization Cabinet with Timer provides a convenient area for setting up PCR reactions in a nucleic acid-free environment.
Brand:  Fisherbrand™ F-UVCABMTY4
1687.29 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15582496
  • 2280.00 € / Each
Explore more special offers
Description
Description
  • UV lights on the cabinets denature nucleic acids in 5 to 30 minutes making them unsuitable for amplification.
  • Incorporates safety features to prevent user exposure to UV light.
  • The UV lights are timer controlled
  • Safety switches on the cabinets doors turn off the UV lights when opened.
  • Shields beta radioactive emissions
  • Specifications
    Specifications
    4 UV Lights, 1 White
    510 mm
    5 x 15 W
    560 mm
    350 mm
    Tray
    12 kg
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Fisherbrand™ Mini UV Sterilization Cabinet with Timer >