missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ MiniCorer

Suitable for sampling sticky, cohesive powders as well as products such as cheese. Sampling Systems™ MiniCorer is excellent at taking samples from semi solid products.

Brand:  Sampling Systems™ 7008A-03

 View more versions of this product

Product Code. 15819391

  • 67.95 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 316L stainless steel
  • High quality bright polish finish
  • Crevice free construction
  • Rotate the handle as the sampler is pushed into the product to allow a core of the product to be cut
  • Remove the sampler when finished
Specifications

Specifications

Corer
Autoclavable
18 mm Max Blade Diameter
Stainless Steel (316L)
Bright
295 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.