Learn More
Lovibond™ Water Testing Minikit
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
Brand:  Lovibond™ 414220
 View more versions of this product
Product Code. 10616151
  • 76.90 € / Each
Explore more special offers
Description
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Specifications
Cyanuric Acid
20 to 200mg/L
Turbidity
Flip top case with calibrated two piece tube, tablet reagents and instructions
Approximately 40 Tests
Minikit water testing kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Water Testing Minikit > Analyte: Cyanuric Acid