missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ ThermaSeal™ 60 Wipers are 100% continuous-filament, double-knit polyester

For ISO Class 3 to 7 (Class 1 to 10,000) applications

Brand:  Texwipe™ TX2069

45.60 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.


Product Code. 15732396

  • 48.00 € / Pack of 150
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 100% continuous-filament double-knit polyester fabric provides good absorbency, wiping efficiency, and abrasion resistance
  • Economical hot-cut wipers polyester for all-purpose cleaning in critical cleanroom applications
  • Thermally sealed edges for fiber control

Order Info

Laundered and packaged in an ISO Class 4 cleanroom using solvent safe Bag-Within-A-Bag™ cleanroom packaging. Basis weight, 122g/m2.

Specifications

Specifications

Dry Wipe
23 cm
23 cm
Autoclavable
100% Continuous filament double-knit construction
9 x 9 in. (22.86 x 22.86 cm)
9 in.
9 in.
Polyester
122
Thermally sealed
For ISO Class 3 to 5 (Class 1 to 100) cleanroom general wiping and cleaning applications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Texwipe™ ThermaSeal™ 60 Wipers are 100% continuous-filament, double-knit polyester > 9 x 9 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.