Learn More
Schmidlin™ Mistral Evolution Nitrogen Generator
This generator produces a continuous supply of high purity nitrogen.
Brand: Schmidlin™ DB-EVO-10-EU
This item is not returnable.
View return policy
Description
- Increases efficiency and return on investment
- On-demand supply 24/7
- Flow rate at 8 barg (116 psig)
- LC/MS grade purity
- Integrated, low noise, oil-free compressors
- Proprietary carbon molecular sieve technology
- Complete 'plug and play' LC/MS solution
Specifications
83.5 cm | |
68 cm | |
3 bar | |
48.5 cm | |
110/120 V at 60 Hz, 220/240 V at 50 Hz | |
1800 W |
10 L/min. | |
35°C | |
19 in. | |
Outlet: 6 mm OD Compression or 1/4 in. | |
>99.5% |