missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ MitoProbe™ JC-1 Assay Kit

MitoProbe™ JC-1 Assay Kit

Brand:  Molecular Probes™ M34152

600.32 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10226762

  • 896.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The MitoProbe™ JC-1 Assay Kit is a quick and reliable assay used to detect changes in mitochondria by flow cytometry.

View a selection guide for all apoptosis assays for flow cytometry.

Selective to mitochondrial changes
The membrane-permeant JC-1 dye is widely used in apoptosis studies to monitor mitochondrial health. JC-1 dye exhibits potential-dependent accumulation in mitochondria, indicated by a fluorescence emission shift from green (∼529 nm) to red (∼590 nm). Consequently, mitochondrial depolarization is indicated by a decrease in the red/green fluorescence intensity ratio. The potential-sensitive color shift is due to concentration-dependent formation of red fluorescent J-aggregates.

Unlike other mitochondrial stains that are affected by plasma membrane potential, the ratio of green to red fluorescence of JC-1 depends only on the membrane potential and not other factors such as mitochondrial size, shape, and density, which may influence single-component fluorescence signals. Use of fluorescence ratio detection allows researchers to make comparative measurements of membrane potential and determine the percentage of mitochondria within a population that respond to an applied stimulus.

Indicator of early apoptosis
A distinctive feature of the early stages of programmed cell death is the disruption of active mitochondria. This mitochondrial disruption includes changes in the membrane potential and alterations to the oxidation-reduction potential of the mitochondria. Changes in the membrane potential are presumed to be due to the opening of the mitochondrial permeability transition pore (MPTP), allowing passage of ions and small molecules. The resulting equilibration of ions leads in turn to the decoupling of the respiratory chain and the release of cytochrome c into the cytosol.

Simple, streamlined protocol
One of the biggest challenges to working with dual-emitting dyes like JC-1 is compensation. The MitoProbe™ JC-1 Assay Kit provides a mitochondrial membrane-potential disrupter, CCCP, in order to produce compensation controls leading to a correctly compensated green-to-red fluorescence ratio. The stain protocol is simple and straightforward: incubate the cells with JC-1 dye and then analyze the fluorescent signal in the FITC and PE channels of of a 488 nm laser.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 5 vials of JC-1 (5′,6,6′-tetrachloro-1,1′,3,3′-tetraethylbenzimidazolylcarbocyanine iodide) (30 μg/vial of powdered dye), 1 vial of DMSO (1.5 mL), 1 vial of CCCP (125 μL , 50 mM in DMSO), and 1 bottle of 10X phosphate-buffered saline (25 mL).

Stored at 2–8°C. Protect from light.
Tube
488
Flow Cytometer
100
MitoProbe™
Room Temperature
Fluorescence
514⁄529, 590
Flow Cytometry
JC-1
JC-1 Assay Kit
1 kit
Mitochondria
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2980661 Certificate TypeCertificate of Analysis Date21/07/2024 Catalog NumberM34152
Lot Number2743038 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberM34152
Lot Number2647633 Certificate TypeCertificate of Analysis Date17/07/2023 Catalog NumberM34152
Lot Number2506011 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog NumberM34152
Lot Number2161890 Certificate TypeCertificate of Analysis Date25/04/2022 Catalog NumberM34152
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ MitoProbe™ JC-1 Assay Kit > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.