Learn More
Invitrogen™ MitoTracker™ Green FM Dye, for flow cytometry
Mitotracker™ dyes for flow cytometry are cell permeant reagents that label mitochondria in live cells. Provided at an optimized concentration with protocols for flow cytometry to allow for more precise analysis with both conventional and spectral flow cytometry.
Brand:  Invitrogen™ M46750
Product Code. 17501655
  • 200.00 € / 100 tests
Edge
Explore more special offers
Description
Description
  • Cell permeant reagents that label mitochondria in live cells
  • Dyes are provided at an optimized concentration with protocols for flow cytometry to allows for more precise analysis with both conventional and spectral flow cytometry
  • Simply incubate cells in the recommended dye concentration for flow cytometry, which passively diffuses across the plasma membrane and accumulates in active mitochondria
  • Bright colors allow for easy detection of mitochondria using flow cytometry without the need for special filters
  • Dyes are consistent, reliable, and stable allowing for the generation of high-quality data
  • Optimized for flow cytometry use with all essential reagents for labeling mitochondria in a single kit
  • Dissolve a vial of dye in anhydrous DMSO to provide enough dye for 20 assays
  • A range of available colors are detected with standard flow cytometry filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry
Dye
5 vials x 20 tests
Room Temperature
5 x 20 tests
100 tests
RUO
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2856022 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberM46750
Lot Number2720276 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog NumberM46750
Lot Number2615847 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberM46750
Lot Number2549289 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog NumberM46750
Lot Number2535872 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog NumberM46750
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ MitoTracker™ Green FM Dye, for flow cytometry > 100 tests